Gene Transcripts

Gene Symbol
SLC13A4
Entrez Gene ID
26266
Gene Name
solute carrier family 13 member 4
Gene Aliases
NAS2, SUT-1, SUT1
Chromosome Location
Chr. 7: 135681237 - 135728204 on Build GRCh38
UniGene ID
Hs.490241
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318192.1 NP_001305121.1 5 1273
NM_012450.3 NP_036582.2 5 1273
XM_011516024.2 XP_011514326.1 5 1053
XM_017011962.1 XP_016867451.1 3 641
XM_017011963.1 XP_016867452.1 5 1053
GenBank mRNA AB208809.1 - 4 1749
AF169301.1 - 5 1243
AK313072.1 - 5 866
BC030689.2 - 5 1255 4821193

View Full Product Details