Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002581.4 | NP_002572.2 | 2 | 1250 | |
XM_017014784.1 | XP_016870273.1 | 2 | 1290 | ||
GenBank mRNA | AK308154.1 | - | 2 | 513 | |
BC078657.1 | - | 2 | 880 | 30340336 | |
X68280.1 | - | 2 | 641 |