Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006231.3 | NP_006222.2 | 3, 4 | 492 | |
XM_011534795.2 | XP_011533097.1 | 3, 4 | 487 | ||
XM_011534799.2 | XP_011533101.1 | 3, 4 | 487 | ||
XM_011534800.2 | XP_011533102.1 | 3, 4 | 487 | ||
GenBank mRNA | BC144561.1 | - | 3, 4 | 315 | |
L09561.1 | - | 3, 4 | 327 | ||
U49356.1 | - | 3, 4 | 283 |