Gene Transcripts

Gene Symbol
UBE2Q2
Entrez Gene ID
92912
Gene Name
ubiquitin conjugating enzyme E2 Q2
Gene Aliases
Chromosome Location
Chr. 15: 75843286 - 75901047 on Build GRCh38
UniGene ID
Hs.23033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145335.1 NP_001138807.1 13 1488
NM_001284382.1 NP_001271311.1 12 1502
NM_173469.3 NP_775740.1 13 1607
XM_005254787.1 XP_005254844.1 10 1167
XM_005254788.1 XP_005254845.1 10 1142
XM_006720770.1 XP_006720833.1 7 930
XM_017022726.1 XP_016878215.1 10 1271
XM_017022727.1 XP_016878216.1 9 1090
XM_017022728.1 XP_016878217.1 9 1065
GenBank mRNA AK000617.1 - 12 1481
AK296234.1 - 13 1488
AL832429.1 - 13 1432
BC006827.1 - 8 670
BC017708.1 - 13 1434 3907760
BC034342.1 - 5 400

View Full Product Details