Gene Transcripts

Gene Symbol
DNAH11
Entrez Gene ID
8701
Gene Name
dynein axonemal heavy chain 11
Gene Aliases
CILD7, DNAHBL, DNAHC11, DNHBL, DPL11
Chromosome Location
Chr. 7: 21543215 - 21901568 on Build GRCh38
UniGene ID
Hs.520245
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277115.1 NP_001264044.1 6 1183
GenBank mRNA AJ320497.1 - 6 1183

View Full Product Details