Gene Transcripts
- Gene Symbol
- CCNC
- Entrez Gene ID
- 892
- Gene Name
- cyclin C
- Gene Aliases
- CycC
- Chromosome Location
- Chr. 6: 99542387 - 99568973 on Build GRCh38
- UniGene ID
- Hs.430646
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001013399.1 |
NP_001013417.1
|
8, 9 |
606 |
|
NM_005190.3 |
NP_005181.2
|
8, 9 |
828 |
|
XM_006715594.1 |
XP_006715657.1
|
8, 9 |
987 |
|
GenBank mRNA |
AB125137.1 |
- |
8, 9 |
615 |
|
AK223247.1 |
- |
8, 9 |
629 |
|
AK291914.1 |
- |
8, 9 |
637 |
|
AK298558.1 |
- |
7, 8 |
567 |
|
BC010135.2 |
- |
8, 9 |
661 |
4106819 |
BC041123.1 |
- |
8, 9 |
811 |
5759974 |
BC056153.1 |
- |
8, 9 |
650 |
6157106 |
M74091.1 |
- |
8, 9 |
629 |
|
View Full Product Details