Gene Transcripts

Gene Symbol
CCNC
Entrez Gene ID
892
Gene Name
cyclin C
Gene Aliases
CycC
Chromosome Location
Chr. 6: 99542387 - 99568973 on Build GRCh38
UniGene ID
Hs.430646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013399.1 NP_001013417.1 12 880
NM_005190.3 NP_005181.2 12 1102
XM_006715594.1 XP_006715657.1 13 1356
GenBank mRNA AB125137.1 - 12 889
AK222768.1 - 12 913
AK223247.1 - 12 903
AK291914.1 - 12 911
AK298558.1 - 11 841
BC010135.2 - 12 935 4106819
BC041123.1 - 12 1085 5759974
BC056153.1 - 12 924 6157106
M74091.1 - 12 903

View Full Product Details