Gene Transcripts

Gene Symbol
LTA4H
Entrez Gene ID
4048
Gene Name
leukotriene A4 hydrolase
Gene Aliases
Chromosome Location
Chr. 12: 96000753 - 96043520 on Build GRCh38
UniGene ID
Hs.524648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000895.2 NP_000886.1 4 577
NM_001256643.1 NP_001243572.1 4 508
NM_001256644.1 NP_001243573.1 4 508
NR_132659.1 4 516
XM_005268871.1 XP_005268928.1 4 567
XM_011538348.1 XP_011536650.1 4 567
XM_011538349.2 XP_011536651.1 4 567
GenBank mRNA AB209740.1 - 4 444
AK293630.1 - 4 496
AK298017.1 - 4 508
AK301308.1 - 4 386
AK308184.1 - 4 506
BC032528.1 - 4 494 5502553
BX647158.1 - 4 465
CR457068.1 - 4 435
DB019628.1 - 4 577
DC358373.1 - 4 509
J02959.1 - 4 465
J03459.1 - 4 503

View Full Product Details