Gene Transcripts

Gene Symbol
SLC25A20
Entrez Gene ID
788
Gene Name
solute carrier family 25 member 20
Gene Aliases
CAC, CACT
Chromosome Location
Chr. 3: 48856923 - 48898993 on Build GRCh38
UniGene ID
Hs.13845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000387.5 NP_000378.1 4 604
GenBank mRNA AK301744.1 - 5 565
AK312962.1 - 4 497
BC001689.2 - 4 476 3050073
BI913588.1 - 4 507
Y10319.1 - 4 467

View Full Product Details