Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167579.1 | NP_001161051.1 | 7 | 774 | |
NM_003052.4 | NP_003043.3 | 7 | 774 | ||
XM_005265975.1 | XP_005266032.1 | 6 | 717 | ||
XM_017009773.1 | XP_016865262.1 | 6 | 717 | ||
XM_017009775.1 | XP_016865264.1 | 6 | 717 | ||
GenBank mRNA | AK096088.1 | - | 7 | 753 | |
AK298299.1 | - | 7 | 771 | ||
BC035294.1 | - | 6 | 874 | ||
BC050385.1 | - | 7 | 751 | 5760855 | |
BC053349.1 | - | 7 | 761 | 5186103 | |
L13258.1 | - | 7 | 751 |