Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024845.2 | NP_001020016.1 | 6 | 886 | |
NM_001261380.1 | NP_001248309.1 | 5 | 833 | ||
NM_006934.3 | NP_008865.2 | 5 | 878 | ||
NM_201649.3 | NP_964012.2 | 6 | 1040 | ||
NR_048548.1 | 5 | 878 | |||
XM_011542017.2 | XP_011540319.1 | 6 | 859 | ||
XM_017002151.1 | XP_016857640.1 | 6 | 794 | ||
XM_017002152.1 | XP_016857641.1 | 5 | 768 | ||
XM_017002153.1 | XP_016857642.1 | 5 | 539 | ||
GenBank mRNA | AK296438.1 | - | 5 | 636 | |
AK298611.1 | - | 4 | 491 | ||
AK304057.1 | - | 6 | 754 | ||
AK311136.1 | - | 6 | 749 | ||
AK315233.1 | - | 5 | 881 | ||
AK315933.1 | - | 5 | 699 | ||
S70609.1 | - | 6 | 1024 | ||
S70612.1 | - | 5 | 862 |