Gene Transcripts

Gene Symbol
PRM1
Entrez Gene ID
5619
Gene Name
protamine 1
Gene Aliases
CT94.1, P1
Chromosome Location
Chr. 16: 11280836 - 11281335 on Build GRCh38
UniGene ID
Hs.2909
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002761.2 NP_002752.1 2 364
GenBank mRNA AY651260.1 - 2 345
CD360017.1 - 2 338
Y00443.1 - 2 367

View Full Product Details