Gene Transcripts
- Gene Symbol
- METTL5
- Entrez Gene ID
- 29081
- Gene Name
- methyltransferase like 5
- Gene Aliases
- HSPC133
- Chromosome Location
- Chr. 2: 169811757 - 169824931 on Build GRCh38
- UniGene ID
- Hs.470553
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293186.1 |
NP_001280115.1
|
4, 5 |
593 |
|
NM_001293187.1 |
NP_001280116.1
|
4, 5 |
507 |
|
NM_014168.3 |
NP_054887.2
|
3, 4 |
734 |
|
XM_011511023.1 |
XP_011509325.1
|
4, 5 |
534 |
|
GenBank mRNA |
AF161482.1 |
- |
4, 5 |
480 |
|
AF201938.1 |
- |
4, 5 |
551 |
|
AK001321.1 |
- |
4, 5 |
481 |
|
AL523293.3 |
- |
3, 4 |
716 |
|
BC000921.2 |
- |
3, 4 |
646 |
3445582 |
BC093014.1 |
- |
4, 5 |
489 |
4799279 |
View Full Product Details