Gene Transcripts
- Gene Symbol
- OSBPL3
- Entrez Gene ID
- 26031
- Gene Name
- oxysterol binding protein like 3
- Gene Aliases
- ORP-3, ORP3, OSBP3
- Chromosome Location
- Chr. 7: 24796537 - 24980218 on Build GRCh38
- UniGene ID
- Hs.520259
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015550.3 |
NP_056365.1
|
9, 10 |
1354 |
|
NM_145320.2 |
NP_663160.1
|
7, 8 |
934 |
|
NM_145321.2 |
NP_663161.1
|
8, 9 |
1027 |
|
NM_145322.2 |
NP_663162.1
|
7, 8 |
934 |
|
NR_104111.1 |
|
8, 9 |
1027 |
|
NR_104112.1 |
|
7, 8 |
934 |
|
XM_005249698.3 |
XP_005249755.1
|
9, 10 |
1074 |
|
XM_006715681.3 |
XP_006715744.1
|
8, 9 |
1248 |
|
XM_006715682.3 |
XP_006715745.1
|
9, 10 |
1360 |
|
XM_006715683.3 |
XP_006715746.1
|
8, 9 |
1267 |
|
XM_006715684.3 |
XP_006715747.1
|
8, 9 |
1027 |
|
XM_011515258.2 |
XP_011513560.1
|
9, 10 |
1151 |
|
XM_011515259.2 |
XP_011513561.1
|
10, 11 |
1648 |
|
XM_017011948.1 |
XP_016867437.1
|
11, 12 |
1511 |
|
XM_017011949.1 |
XP_016867438.1
|
10, 11 |
1648 |
|
GenBank mRNA |
AB014604.2 |
- |
9, 10 |
1291 |
|
AF392444.1 |
- |
9, 10 |
1263 |
|
AF491781.1 |
- |
8, 9 |
981 |
|
AF491782.1 |
- |
7, 8 |
888 |
|
AF491783.1 |
- |
8, 9 |
981 |
|
AF491784.1 |
- |
7, 8 |
888 |
|
AF491785.1 |
- |
8, 9 |
981 |
|
AF491786.1 |
- |
7, 8 |
888 |
|
AF515639.1 |
- |
2, 3 |
141 |
|
AF515640.1 |
- |
1, 2 |
48 |
|
AL832293.1 |
- |
6, 7 |
590 |
|
AY008372.1 |
- |
9, 10 |
1184 |
|
BC017731.1 |
- |
9, 10 |
1283 |
3909164 |
View Full Product Details