Gene Transcripts

Gene Symbol
SIDT2
Entrez Gene ID
51092
Gene Name
SID1 transmembrane family member 2
Gene Aliases
CGI-40
Chromosome Location
Chr. 11: 117179223 - 117197445 on Build GRCh38
UniGene ID
Hs.712144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040455.1 NP_001035545.1 11 1114
GenBank mRNA AF151799.1 - 11 1156
AK075137.1 - 2 92
AK292501.1 - 11 1594
AK292974.1 - 11 1105
AK301501.1 - 3 384
AK303023.1 - 11 1070
AY358442.1 - 11 1123
BC114522.1 - 12 1155 40035836
BX648437.1 - 3 212
KC109742.1 - 11 1070

View Full Product Details