Gene Transcripts
- Gene Symbol
- PSMD13
- Entrez Gene ID
- 5719
- Gene Name
- proteasome 26S subunit, non-ATPase 13
- Gene Aliases
- HSPC027, Rpn9, S11, p40.5
- Chromosome Location
- Chr. 11: 236675 - 252984 on Build GRCh38
- UniGene ID
- Hs.134688
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002817.3 |
NP_002808.3
|
6, 7 |
637 |
|
NM_175932.2 |
NP_787128.2
|
4, 5 |
643 |
|
XM_011520235.2 |
XP_011518537.1
|
6, 7 |
770 |
|
GenBank mRNA |
AB009398.1 |
- |
6, 7 |
464 |
|
AF083245.1 |
- |
6, 7 |
449 |
|
AF086708.1 |
- |
6, 7 |
395 |
|
AF107837.1 |
- |
6, 7 |
428 |
|
AK094775.1 |
- |
4, 5 |
474 |
|
AK222895.1 |
- |
6, 7 |
468 |
|
AK295944.1 |
- |
5, 6 |
434 |
|
AK307447.1 |
- |
5, 6 |
362 |
|
AK314733.1 |
- |
6, 7 |
899 |
|
BC001100.2 |
- |
6, 7 |
450 |
3506530 |
BC001747.1 |
- |
6, 7 |
418 |
3535807 |
BM456586.1 |
- |
6, 7 |
468 |
|
BT007307.1 |
- |
6, 7 |
395 |
|
BX647314.1 |
- |
5, 6 |
372 |
|
View Full Product Details