Gene Transcripts

Gene Symbol
SLC25A33
Entrez Gene ID
84275
Gene Name
solute carrier family 25 member 33
Gene Aliases
BMSC-MCP, PNC1
Chromosome Location
Chr. 1: 9539469 - 9582773 on Build GRCh38
UniGene ID
Hs.568613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032315.2 NP_115691.1 7 1423
XM_005263503.3 XP_005263560.1 7 1092
XM_011542296.1 XP_011540598.1 7 1210
GenBank mRNA AJ880283.1 - 7 1409
BC004991.1 - 7 1392 2905957
BC073135.1 - 7 1409 6172562

View Full Product Details