Gene Transcripts

Gene Symbol
RBMS2
Entrez Gene ID
5939
Gene Name
RNA binding motif single stranded interacting protein 2
Gene Aliases
SCR3
Chromosome Location
Chr. 12: 56521812 - 56596196 on Build GRCh38
UniGene ID
Hs.505729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002898.3 NP_002889.1 3, 4 499
XM_005269059.4 XP_005269116.1 3, 4 367
XM_005269060.4 XP_005269117.1 3, 4 367
XM_005269061.3 XP_005269118.1 3, 4 367
XM_005269066.4 XP_005269123.1 3, 4 367
XM_006719541.3 XP_006719604.1 3, 4 367
XM_006719542.3 XP_006719605.1 3, 4 367
XM_006719543.3 XP_006719606.1 3, 4 367
XM_006719544.3 XP_006719607.1 3, 4 367
XM_011538637.2 XP_011536939.1 3, 4 311
XM_011538638.2 XP_011536940.1 4, 5 633
XM_011538639.2 XP_011536941.1 3, 4 367
XM_011538640.2 XP_011536942.1 3, 4 367
XM_017019775.1 XP_016875264.1 3, 4 306
GenBank mRNA AK293315.1 - 2, 3 159
AK299580.1 - 3, 4 315
AK304540.1 - 2, 3 159
BC027863.1 - 3, 4 308 5202634
BC072679.1 - 3, 4 372 6048623
D28483.1 - 3, 4 316

View Full Product Details