Gene Transcripts

Gene Symbol
PLD5
Entrez Gene ID
200150
Gene Name
phospholipase D family member 5
Gene Aliases
PLDC
Chromosome Location
Chr. 1: 242085051 - 242530546 on Build GRCh38
UniGene ID
Hs.672452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195811.1 NP_001182740.1 6, 7 870
NM_001195812.1 NP_001182741.1 5, 6 708
NM_001320272.1 NP_001307201.1 7, 8 1164
NM_152666.2 NP_689879.2 7, 8 1157
XM_006711752.2 XP_006711815.1 6, 7 1324
XM_011544115.2 XP_011542417.1 8, 9 1232
XM_011544116.2 XP_011542418.1 8, 9 1060
XM_011544119.2 XP_011542421.2 3, 4 1231
XM_011544120.2 XP_011542422.1 6, 7 876
XM_011544121.2 XP_011542423.1 3, 4 365
XM_011544122.2 XP_011542424.1 3, 4 1826
XM_017000567.1 XP_016856056.1 6, 7 1337
XM_017000568.1 XP_016856057.1 6, 7 858
XM_017000569.1 XP_016856058.1 6, 7 2837
XM_017000570.1 XP_016856059.1 4, 5 1577
GenBank mRNA AK091691.1 - 5, 6 682
AK098092.1 - 7, 8 1164
AK295421.1 - 6, 7 851
AK307857.1 - 6, 7 849
AY391836.1 - 5, 6 720
AY461578.1 - 6, 7 870
BC101373.1 - 6, 7 797 40025860
BC101374.1 - 6, 7 797 40025861
BC101375.1 - 6, 7 797 40025862

View Full Product Details