Gene Transcripts

Gene Symbol
RILPL1
Entrez Gene ID
353116
Gene Name
Rab interacting lysosomal protein like 1
Gene Aliases
GOSPEL, RLP1
Chromosome Location
Chr. 12: 123471073 - 123533718 on Build GRCh38
UniGene ID
Hs.530315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319243.1 NP_001306172.1 6, 7 1306
GenBank mRNA AK096697.1 - 7, 8 1148

View Full Product Details