Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168278.1 | NP_001161750.1 | 8 | 1458 | |
NM_001168280.1 | NP_001161752.1 | 7 | 1405 | ||
NM_015472.4 | NP_056287.1 | 7 | 1541 | ||
XM_017006121.1 | XP_016861610.1 | 10 | 1651 | ||
XM_017006122.1 | XP_016861611.1 | 8 | 1450 | ||
GenBank mRNA | AJ299431.1 | - | 7 | 1376 | |
AK022036.1 | - | 8 | 1458 | ||
AL050107.1 | - | 5 | 765 | ||
AL833852.1 | - | 6 | 1195 | ||
BC014052.2 | - | 7 | 1435 | 3635491 |