Gene Transcripts
- Gene Symbol
- MARK1
- Entrez Gene ID
- 4139
- Gene Name
- microtubule affinity regulating kinase 1
- Gene Aliases
- MARK, Par-1c, Par1c
- Chromosome Location
- Chr. 1: 220528183 - 220664457 on Build GRCh38
- UniGene ID
- Hs.497806
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286124.1 |
NP_001273053.1
|
7, 8 |
1200 |
|
NM_001286126.1 |
NP_001273055.1
|
7, 8 |
1200 |
|
NM_001286128.1 |
NP_001273057.1
|
6, 7 |
1134 |
|
NM_018650.4 |
NP_061120.3
|
7, 8 |
1200 |
|
XM_005273134.4 |
XP_005273191.1
|
7, 8 |
1067 |
|
XM_006711326.3 |
XP_006711389.1
|
7, 8 |
461 |
|
XM_011509561.2 |
XP_011507863.1
|
7, 8 |
1196 |
|
XM_011509562.2 |
XP_011507864.1
|
7, 8 |
813 |
|
XM_017001305.1 |
XP_016856794.1
|
8, 9 |
657 |
|
XM_017001306.1 |
XP_016856795.1
|
7, 8 |
697 |
|
XM_017001307.1 |
XP_016856796.1
|
8, 9 |
1097 |
|
GenBank mRNA |
AB040910.1 |
- |
6, 7 |
1091 |
|
AF154845.1 |
- |
7, 8 |
826 |
|
AK027493.1 |
- |
3, 4 |
157 |
|
AK295506.1 |
- |
7, 8 |
698 |
|
AK314614.1 |
- |
7, 8 |
860 |
|
BC113869.1 |
- |
7, 8 |
736 |
40036731 |
BC114478.1 |
- |
7, 8 |
736 |
40036730 |
KJ534879.1 |
- |
7, 8 |
560 |
|
KJ534881.1 |
- |
7, 8 |
560 |
|
KJ535089.1 |
- |
7, 8 |
559 |
|
View Full Product Details