Gene Transcripts
- Gene Symbol
- TIMP1
- Entrez Gene ID
- 7076
- Gene Name
- TIMP metallopeptidase inhibitor 1
- Gene Aliases
- CLGI, EPA, EPO, HCI, TIMP, TIMP-1
- Chromosome Location
- Chr. X: 47582291 - 47586791 on Build GRCh38
- UniGene ID
- Hs.522632
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003254.2 |
NP_003245.1
|
4, 5 |
512 |
|
GenBank mRNA |
A10416.1 |
- |
4, 5 |
382 |
|
AK074854.1 |
- |
4, 5 |
490 |
|
AK311937.1 |
- |
4, 5 |
364 |
|
BC000866.1 |
- |
4, 5 |
465 |
3460153 |
BC007097.1 |
- |
4, 5 |
413 |
|
BU857950.1 |
- |
|
559 |
|
CR407638.1 |
- |
3, 4 |
320 |
|
CR541982.1 |
- |
3, 4 |
320 |
|
M12670.1 |
- |
4, 5 |
368 |
|
M59906.1 |
- |
4, 5 |
363 |
|
S68252.1 |
- |
3, 4 |
323 |
|
X02598.1 |
- |
4, 5 |
361 |
|
X03124.1 |
- |
4, 5 |
382 |
|
View Full Product Details