Gene Transcripts
- Gene Symbol
- TRIM43
- Entrez Gene ID
- 129868
- Gene Name
- tripartite motif containing 43
- Gene Aliases
- TRIM43A
- Chromosome Location
- Chr. 2: 95592018 - 95599723 on Build GRCh38
- UniGene ID
- Hs.232026
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138800.1 |
NP_620155.1
|
6, 7 |
993 |
|
GenBank mRNA |
BC015353.1 |
- |
6, 7 |
993 |
|
- Gene Symbol
- TRIM43B
- Entrez Gene ID
- 653192
- Gene Name
- tripartite motif containing 43B
- Gene Aliases
- Chromosome Location
- Chr. 2: 95476967 - 95484731 on Build GRCh38
- UniGene ID
- Hs.723811
- Species
- Homo sapiens
View Full Product Details