Gene Transcripts

Gene Symbol
TDRD5
Entrez Gene ID
163589
Gene Name
tudor domain containing 5
Gene Aliases
TUDOR3
Chromosome Location
Chr. 1: 179591613 - 179691272 on Build GRCh38
UniGene ID
Hs.197354
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199085.1 NP_001186014.1 3 813
NM_001199089.1 NP_001186018.1 3 526
NM_001199091.1 NP_001186020.1 3 654
NM_173533.3 NP_775804.2 3 813
XM_005244934.1 XP_005244991.1 3 598
XM_005244935.4 XP_005244992.1 3 503
XM_017000473.1 XP_016855962.1 3 519
GenBank mRNA AK126889.1 - 3 653
AK127370.1 - 3 545
BC130532.1 - 3 490 40146563
BC144058.1 - 3 545 9052578

View Full Product Details