Gene Transcripts

Gene Symbol
RCOR2
Entrez Gene ID
283248
Gene Name
REST corepressor 2
Gene Aliases
Chromosome Location
Chr. 11: 63911221 - 63916844 on Build GRCh38
UniGene ID
Hs.98788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173587.3 NP_775858.2 9 1335
XM_005273932.3 XP_005273989.1 9 1052
GenBank mRNA AB593132.1 - 9 1678
BC010608.1 - 2 293
BC023587.2 - 9 1335 4762190

View Full Product Details