Gene Transcripts

Gene Symbol
TAS2R41
Entrez Gene ID
259287
Gene Name
taste 2 receptor member 41
Gene Aliases
T2R41, T2R59
Chromosome Location
Chr. 7: 143477873 - 143478796 on Build GRCh38
UniGene ID
Hs.650648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176883.2 NP_795364.2 1 631
GenBank mRNA BC101157.3 - 1 631
BC101158.2 - 1 631 40015713

View Full Product Details