Gene Transcripts

Gene Symbol
KAZALD1
Entrez Gene ID
81621
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
BONO1, FKSG28, FKSG40, IGFBP-rP10
Chromosome Location
Chr. 10: 101061141 - 101068141 on Build GRCh38
UniGene ID
Hs.534859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030929.4 NP_112191.2 5 2483
GenBank mRNA AK172862.1 - 5 1734
BC007758.2 - 5 1884 4122366
DA422973.1 - 1 368

View Full Product Details