Gene Transcripts

Gene Symbol
PCDHB4
Entrez Gene ID
56131
Gene Name
protocadherin beta 4
Gene Aliases
PCDH-BETA4
Chromosome Location
Chr. 5: 141121750 - 141125623 on Build GRCh38
UniGene ID
Hs.591250
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018938.3 NP_061761.1 1 3242
GenBank mRNA AF217754.1 - 1 2993

View Full Product Details