Gene Transcripts

Gene Symbol
HRH4
Entrez Gene ID
59340
Gene Name
histamine receptor H4
Gene Aliases
AXOR35, BG26, GPCR105, GPRv53, H4, H4R, HH4R
Chromosome Location
Chr. 18: 24460568 - 24487427 on Build GRCh38
UniGene ID
Hs.287388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143828.1 NP_001137300.1 2 605
NM_001160166.1 NP_001153638.1 2 705
NM_021624.3 NP_067637.2 3 869
GenBank mRNA AB044934.1 - 3 823
AB045370.1 - 3 832
AF307973.1 - 3 769
AF312230.1 - 3 869
AF325356.1 - 3 769
AF329449.1 - 3 769
AJ298292.1 - 3 769
AY008280.1 - 3 769
AY136745.1 - 3 769
BC069136.1 - 3 869 7216992
BC112348.1 - 3 829 40006540
DQ835186.1 - 2 505

View Full Product Details