Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012158.2 | NP_036290.1 | 5 | 1232 | |
XM_005266336.1 | XP_005266393.1 | 5 | 1015 | ||
XM_005266337.2 | XP_005266394.1 | 4 | 867 | ||
GenBank mRNA | AF126028.1 | - | 4 | 836 | |
AF129532.1 | - | 5 | 899 | ||
AK025484.1 | - | 3 | 430 | ||
AK314442.1 | - | 5 | 1200 | ||
AL833187.1 | - | 5 | 1180 | ||
BC072448.1 | - | 5 | 1186 | 6166244 |