Gene Transcripts

Gene Symbol
L3MBTL1
Entrez Gene ID
26013
Gene Name
l(3)mbt-like 1 (Drosophila)
Gene Aliases
H-L(3)MBT, L3MBTL, ZC2HC3, dJ138B7.3
Chromosome Location
Chr. 20: 43507680 - 43541895 on Build GRCh38
UniGene ID
Hs.709356
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015478.6 NP_056293.4 19 2261
NM_032107.4 NP_115479.4 22 2432
GenBank mRNA AK123176.1 - 3 1414
AK299199.1 - 22 2402
AL110279.1 - 10 3071
BC039820.1 - 19 2176 5199689

View Full Product Details