Gene Transcripts
- Gene Symbol
- RNASET2
- Entrez Gene ID
- 8635
- Gene Name
- ribonuclease T2
- Gene Aliases
- RNASE6PL, bA514O12.3
- Chromosome Location
- Chr. 6: 166929509 - 166956589 on Build GRCh38
- UniGene ID
- Hs.529989
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003730.4 |
NP_003721.2
|
8, 9 |
982 |
|
XM_017011397.1 |
XP_016866886.1
|
9, 10 |
761 |
|
XM_017011398.1 |
XP_016866887.1
|
7, 8 |
883 |
|
XM_017011399.1 |
XP_016866888.1
|
7, 8 |
897 |
|
GenBank mRNA |
AJ419865.1 |
- |
8, 9 |
936 |
|
AJ419866.1 |
- |
8, 9 |
1656 |
|
AK124363.1 |
- |
1, 2 |
2014 |
|
AK315467.1 |
- |
8, 9 |
943 |
|
BC001660.2 |
- |
8, 9 |
908 |
2959583 |
BC001819.2 |
- |
8, 9 |
908 |
2959583 |
BC036282.1 |
- |
8, 9 |
969 |
|
BC039713.1 |
- |
8, 9 |
911 |
5225327 |
BC051912.1 |
- |
8, 9 |
921 |
5227463 |
U85625.2 |
- |
8, 9 |
939 |
|
View Full Product Details