Gene Transcripts

Gene Symbol
PIP5K1C
Entrez Gene ID
23396
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 gamma
Gene Aliases
LCCS3, PIP5K-GAMMA, PIP5K1-gamma, PIP5Kgamma
Chromosome Location
Chr. 19: 3630181 - 3700492 on Build GRCh38
UniGene ID
Hs.282177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195733.1 NP_001182662.1 17 2294
NM_012398.2 NP_036530.1 18 2378
XM_017026540.1 XP_016882029.1 17 2352
GenBank mRNA AB011161.1 - 18 2346

View Full Product Details