Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003803.3 | NP_003794.3 | 20 | 3419 | |
NM_019856.1 | NP_062830.1 | 19 | 3131 | ||
XM_017026062.1 | XP_016881551.1 | 20 | 3146 | ||
GenBank mRNA | AJ621424.1 | - | 20 | 3419 | |
AK125322.1 | - | 21 | 3419 | ||
BC116183.1 | - | 19 | 2872 | 40073728 | |
X69090.1 | - | 17 | 2503 |