Gene Transcripts
- Gene Symbol
- EIF3B
- Entrez Gene ID
- 8662
- Gene Name
- eukaryotic translation initiation factor 3 subunit B
- Gene Aliases
- EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, PRT1
- Chromosome Location
- Chr. 7: 2354540 - 2380745 on Build GRCh38
- UniGene ID
- Hs.371001
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001037283.1 |
NP_001032360.1
|
10, 11 |
1705 |
|
NM_003751.3 |
NP_003742.2
|
10, 11 |
1705 |
|
XM_011515599.1 |
XP_011513901.1
|
10, 11 |
2004 |
|
XM_011515600.1 |
XP_011513902.1
|
10, 11 |
2004 |
|
XM_011515601.1 |
XP_011513903.1
|
11, 12 |
1887 |
|
XM_017012752.1 |
XP_016868241.1
|
10, 11 |
2004 |
|
GenBank mRNA |
AB209381.1 |
- |
3, 4 |
3255 |
|
AK300967.1 |
- |
11, 12 |
1447 |
|
AK302058.1 |
- |
11, 12 |
1231 |
|
BC001173.1 |
- |
10, 11 |
1653 |
3501759 |
BC009986.1 |
- |
9, 10 |
1047 |
|
BC110865.1 |
- |
10, 11 |
1631 |
6175284 |
U62583.1 |
- |
10, 11 |
1675 |
|
U78525.1 |
- |
10, 11 |
1697 |
|
View Full Product Details