Gene Transcripts

Gene Symbol
WNT8B
Entrez Gene ID
7479
Gene Name
Wnt family member 8B
Gene Aliases
Chromosome Location
Chr. 10: 100463055 - 100483642 on Build GRCh38
UniGene ID
Hs.421281
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003393.3 NP_003384.2 6 1933
GenBank mRNA AB073637.1 - 6 1931
Y11094.1 - 6 1954

View Full Product Details