Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256312.1 | NP_001243241.1 | 5 | 622 | |
NM_001256313.1 | NP_001243242.1 | 3 | 269 | ||
NM_006281.3 | NP_006272.2 | 3 | 279 | ||
XM_011517248.2 | XP_011515550.2 | 4 | 2252 | ||
XM_011517251.1 | XP_011515553.1 | 3 | 293 | ||
XM_011517252.2 | XP_011515554.2 | 4 | 2252 | ||
XM_011517257.2 | XP_011515559.2 | 4 | 2252 | ||
XM_017013756.1 | XP_016869245.1 | 4 | 2252 | ||
XM_017013757.1 | XP_016869246.1 | 4 | 2252 | ||
XM_017013758.1 | XP_016869247.1 | 3 | 293 | ||
XM_017013759.1 | XP_016869248.1 | 3 | 311 | ||
XM_017013760.1 | XP_016869249.1 | 3 | 264 | ||
XM_017013761.1 | XP_016869250.1 | 3 | 293 | ||
XM_017013762.1 | XP_016869251.1 | 4 | 2252 | ||
GenBank mRNA | AK075229.1 | - | 3 | 265 | |
AK131363.1 | - | 5 | 622 | ||
AK291837.1 | - | 3 | 243 | ||
AK307787.1 | - | 3 | 269 | ||
AK312660.1 | - | 3 | 272 | ||
BC010640.2 | - | 3 | 268 | 3868822 | |
U26424.1 | - | 3 | 276 | ||
U60206.1 | - | 3 | 213 |