Gene Transcripts

Gene Symbol
SHOC2
Entrez Gene ID
8036
Gene Name
SHOC2, leucine rich repeat scaffold protein
Gene Aliases
SIAA0862, SOC2, SUR8
Chromosome Location
Chr. 10: 110919370 - 111013667 on Build GRCh38
UniGene ID
Hs.104315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001269039.2 NP_001255968.1 4, 5 1380
NM_001324336.1 NP_001311265.1 5, 6 1475
NM_001324337.1 NP_001311266.1 6, 7 1703
NM_007373.3 NP_031399.2 5, 6 1518
NR_136749.1 4, 5 581
XM_017016702.1 XP_016872191.1 5, 6 1423
XM_017016703.1 XP_016872192.1 5, 6 7240
XM_017016704.1 XP_016872193.1 5, 6 1422
GenBank mRNA AB020669.1 - 5, 6 1446
AF054828.1 - 5, 6 1440
AF068920.1 - 4, 5 1169
AK090820.1 - 3, 4 539
AK292833.1 - 6, 7 1644
AK293271.1 - 4, 5 520
BC044752.1 - 5, 6 1454
BC050445.1 - 4, 5 1373 6067669
BX649055.1 - 6, 7 1286

View Full Product Details