Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001303627.1 | NP_001290556.1 | 2, 3 | 312 | |
NM_014730.3 | NP_055545.1 | 2, 3 | 595 | ||
XM_011539031.1 | XP_011537333.1 | 2, 3 | 5010 | ||
XM_011539032.1 | XP_011537334.1 | 3, 4 | 4698 | ||
GenBank mRNA | BC000371.2 | - | 2, 3 | 544 | |
BC016297.2 | - | 2, 3 | 546 | 3832682 | |
BI458950.1 | - | 2, 3 | 314 | ||
BU845446.1 | - | 573 | |||
D63486.1 | - | 2, 3 | 550 |