Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022841.5 | NP_073752.5 | 9 | 1952 | |
XM_005254603.3 | XP_005254660.2 | 10 | 2046 | ||
XM_011521925.2 | XP_011520227.1 | 9 | 1884 | ||
XM_017022506.1 | XP_016877995.1 | 10 | 4769 | ||
XM_017022507.1 | XP_016877996.1 | 9 | 2012 | ||
XM_017022508.1 | XP_016877997.1 | 8 | 3181 | ||
GenBank mRNA | AK023056.1 | - | 1 | 247 | |
AK126103.1 | - | 2 | 1104 | ||
AK128045.1 | - | 9 | 1889 | ||
AL832584.1 | - | 1 | 762 | ||
AL833367.1 | - | 1 | 179 | ||
BC112936.1 | - | 7 | 1772 | 6014850 | |
BC143438.1 | - | 7 | 1709 |