Gene Transcripts

Gene Symbol
SRRM4
Entrez Gene ID
84530
Gene Name
serine/arginine repetitive matrix 4
Gene Aliases
KIAA1853, MU-MB-2.76, nSR100
Chromosome Location
Chr. 12: 118981495 - 119163051 on Build GRCh38
UniGene ID
Hs.744964
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194286.3 NP_919262.2 13 6183
GenBank mRNA AK125400.1 - 1 922

View Full Product Details