Gene Transcripts

Gene Symbol
PSME2
Entrez Gene ID
5721
Gene Name
proteasome activator subunit 2
Gene Aliases
PA28B, PA28beta, REGbeta
Chromosome Location
Chr. 14: 24143365 - 24147316 on Build GRCh38
UniGene ID
Hs.434081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002818.2 NP_002809.2 9, 10 613
XM_006720213.1 XP_006720276.1 9, 10 1407
GenBank mRNA AK026580.1 - 9, 10 634
AK225876.1 - 9, 10 576
BC004368.1 - 9, 10 563 3640584
BC019885.1 - 9, 10 563 4994572
BC072025.1 - 9, 10 562 6270762
BX161498.1 - 9, 10 1188
CR541657.1 - 9, 10 548
CR541743.1 - 9, 10 548
D45248.1 - 9, 10 613

View Full Product Details