Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006267.4 | NP_006258.3 | 20 | 2870 | |
XM_005264002.2 | XP_005264059.1 | 20 | 2905 | ||
XM_005264003.2 | XP_005264060.1 | 20 | 2833 | ||
XM_005264004.2 | XP_005264061.1 | 20 | 2833 | ||
XM_005264005.4 | XP_005264062.1 | 20 | 2893 | ||
XM_011511575.2 | XP_011509877.1 | 20 | 2808 | ||
XM_011511576.2 | XP_011509878.1 | 20 | 2833 | ||
XM_017004623.1 | XP_016860112.1 | 20 | 2833 | ||
XM_017004624.1 | XP_016860113.1 | 19 | 2738 | ||
GenBank mRNA | AB209483.1 | - | 17 | 2486 | |
AK026993.1 | - | 4 | 299 | ||
D42063.1 | - | 20 | 2871 |