Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004148.3 | NP_004139.2 | 4 | 1152 | |
XM_011518716.1 | XP_011517018.1 | 5 | 1225 | ||
GenBank mRNA | AK094530.1 | - | 2 | 1908 | |
BC000298.1 | - | 4 | 1117 | 2820092 | |
BC004440.1 | - | 4 | 1108 | 2820092 | |
BC019336.1 | - | 4 | 1107 | 3617778 | |
BC048212.1 | - | 4 | 1152 | 5806415 | |
U72661.1 | - | 4 | 1101 | ||
U91512.1 | - | 4 | 1101 |