Gene Transcripts

Gene Symbol
ROR2
Entrez Gene ID
4920
Gene Name
receptor tyrosine kinase like orphan receptor 2
Gene Aliases
BDB, BDB1, NTRKR2
Chromosome Location
Chr. 9: 91722596 - 91950206 on Build GRCh38
UniGene ID
Hs.644776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318204.1 NP_001305133.1 3 620
NM_004560.3 NP_004551.2 3 620
XM_005252008.4 XP_005252065.1 3 585
XM_006717121.3 XP_006717184.1 3 1242
XM_017014762.1 XP_016870251.1 3 2317
XM_017014763.1 XP_016870252.1 3 496
GenBank mRNA AB209154.1 - 5 889
BC033697.1 - 3 607
BC051273.1 - 3 432
BC130522.1 - 3 508 40146553
M97639.1 - 3 620

View Full Product Details