Gene Transcripts
- Gene Symbol
- PEG3
- Entrez Gene ID
- 5178
- Gene Name
- paternally expressed 3
- Gene Aliases
- PW1, ZKSCAN22, ZNF904, ZSCAN24
- Chromosome Location
- Chr. 19: 56810077 - 56840726 on Build GRCh38
- UniGene ID
- Hs.731875
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146184.1 |
NP_001139656.1
|
8, 9 |
1177 |
|
NM_001146185.1 |
NP_001139657.1
|
8, 9 |
863 |
|
NM_001146186.1 |
NP_001139658.1
|
6, 7 |
1210 |
|
NM_001146187.1 |
NP_001139659.1
|
8, 9 |
869 |
|
NM_006210.2 |
NP_006201.1
|
9, 10 |
1253 |
|
GenBank mRNA |
AB006625.2 |
- |
8, 9 |
842 |
|
AF208967.1 |
- |
9, 10 |
1223 |
|
AF208968.1 |
- |
8, 9 |
1147 |
|
AF208969.1 |
- |
8, 9 |
833 |
|
AF208970.1 |
- |
6, 7 |
1211 |
|
AK295679.1 |
- |
8, 9 |
869 |
|
AK307476.1 |
- |
8, 9 |
869 |
|
BC136268.1 |
- |
7, 8 |
697 |
9020255 |
BC150272.1 |
- |
8, 9 |
842 |
9007244 |
View Full Product Details