Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002126.4 | NP_002117.1 | 4 | 1784 | |
XM_005257269.2 | XP_005257326.1 | 4 | 1767 | ||
XM_011524705.1 | XP_011523007.1 | 4 | 1241 | ||
XM_017024556.1 | XP_016880045.1 | 4 | 1238 | ||
GenBank mRNA | AK074289.1 | - | 3 | 1248 | |
AK290043.1 | - | 4 | 1221 | ||
AK295726.1 | - | 3 | 1276 | ||
BC036093.1 | - | 4 | 1738 | 5288156 | |
M95585.1 | - | 4 | 1581 |