Gene Transcripts

Gene Symbol
ANXA11
Entrez Gene ID
311
Gene Name
annexin A11
Gene Aliases
ANX11, CAP50
Chromosome Location
Chr. 10: 80153953 - 80205677 on Build GRCh38
UniGene ID
Hs.530291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001157.2 NP_001148.1 6, 7 977
NM_001278407.1 NP_001265336.1 8, 9 956
NM_001278408.1 NP_001265337.1 8, 9 1564
NM_001278409.1 NP_001265338.1 8, 9 1295
NM_145868.1 NP_665875.1 7, 8 1026
NM_145869.1 NP_665876.1 8, 9 1222
XM_005269741.4 XP_005269798.1 4, 5 1054
XM_005269742.1 XP_005269799.1 6, 7 980
XM_006717813.1 XP_006717876.1 7, 8 991
XM_006717814.3 XP_006717877.1 7, 8 980
XM_011539735.1 XP_011538037.1 8, 9 1505
XM_011539736.2 XP_011538038.1 8, 9 1505
GenBank mRNA AB209770.1 - 7, 8 971
AJ278463.1 - 6, 7 976
AJ278464.1 - 7, 8 1025
AJ278465.1 - 8, 9 1221
AK000567.1 - 6, 7 976
AK056107.1 - 8, 9 956
AK126287.1 - 8, 9 1564
AK222918.1 - 6, 7 939
AK298362.1 - 7, 8 787
AK300509.1 - 4, 5 1089
AK301047.1 - 8, 9 1280
BC007564.2 - 7, 8 812 2989058
BT019934.1 - 5, 6 739
CR450323.1 - 5, 6 739
L19605.1 - 6, 7 917

View Full Product Details