Gene Transcripts

Gene Symbol
PLK2
Entrez Gene ID
10769
Gene Name
polo like kinase 2
Gene Aliases
SNK, hPlk2, hSNK
Chromosome Location
Chr. 5: 58453982 - 58460139 on Build GRCh38
UniGene ID
Hs.398157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252226.1 NP_001239155.1 3 493
NM_006622.3 NP_006613.2 2 535
GenBank mRNA AF059617.1 - 2 655
AF223574.1 - 2 478
AK027851.1 - 2 480
AK098163.1 - 3 461
AK297298.1 - 3 438
AK304746.1 - 1 355
AK312505.1 - 2 483
BC013879.2 - 2 468 3831747
DA020501.1 - 2 482
U85755.1 - 1 30

View Full Product Details